Assay Details
| Gene Symbol: ZFP36L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639346.4 | XP_003639394.1 | 1 - 2 | 365 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZFP36L2| Gene Symbol: ZFP36L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639346.4 | XP_003639394.1 | 1 - 2 | 365 | 60 | |
Set Membership: |
|