Assay Details
| Gene Symbol: NAA60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621621.3 | XP_005621678.1 | 3 - 4 | 916 | 85 | |
| XM_014114663.2 | XP_013970138.1 | 2 - 3 | 386 | 85 | ||
| XM_014114664.2 | XP_013970139.1 | 2 - 3 | 285 | 85 | ||
| XM_846749.5 | XP_851842.1 | 2 - 3 | 282 | 85 | ||
More Information
Set Membership: |
|

NAA60
Genomic Map