Assay Details
Gene Symbol: DAXX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627201.3 | XP_005627258.1 | 6 - 7 | 2116 | 66 | |
XM_005627204.3 | XP_005627261.1 | 6 - 7 | 2108 | 66 | ||
XM_005627205.3 | XP_005627262.1 | 5 - 6 | 1741 | 66 | ||
XM_022425929.1 | XP_022281637.1 | 5 - 6 | 1925 | 66 | ||
XM_022425930.1 | XP_022281638.1 | 6 - 7 | 2114 | 66 | ||
XM_022425931.1 | XP_022281639.1 | 6 - 7 | 2112 | 66 | ||
XM_022425932.1 | XP_022281640.1 | 6 - 7 | 2055 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
