Assay Details
| Gene Symbol: HSD17B8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627198.3 | XP_005627255.2 | 8 - 9 | 869 | 55 | |
| XM_014118363.2 | XP_013973838.1 | 7 - 8 | 827 | 55 | ||
| XM_014118364.2 | XP_013973839.1 | 7 - 8 | 805 | 55 | ||
| XM_845911.5 | XP_851004.3 | 8 - 9 | 848 | 55 | ||
More Information
Set Membership: |
|

HSD17B8
Genomic Map