Assay Details
| Gene Symbol: WBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022408369.1 | XP_022264077.1 | 4 - 5 | 421 | 93 | |
| XM_022408370.1 | XP_022264078.1 | 4 - 5 | 420 | 93 | ||
| XM_534136.6 | XP_534136.2 | 4 - 5 | 418 | 93 | ||
More Information
Set Membership: |
|

WBP4
Genomic Map