Assay Details
| Gene Symbol: LOC476900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014106246.2 | XP_013961721.1 | 6 - 7 | 795 | 91 | |
| XM_022407958.1 | XP_022263666.1 | 6 - 7 | 694 | 91 | ||
More Information
Set Membership: |
|

LOC476900
Genomic Map