Assay Details
| Gene Symbol: RFC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410522.1 | XP_022266230.1 | 7 - 8 | 776 | 67 | |
| XM_022410523.1 | XP_022266231.1 | 7 - 8 | 776 | 67 | ||
| XM_534696.6 | XP_534696.3 | 7 - 8 | 774 | 67 | ||
More Information
Set Membership: |
|

RFC5
Genomic Map