Assay Details
| Gene Symbol: LOC608697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014106509.2 | XP_013961984.1 | 3 - 4 | 285 | 69 | |
| XM_014106515.2 | XP_013961990.1 | 3 - 4 | 285 | 69 | ||
| XM_022408603.1 | XP_022264311.1 | 3 - 4 | 285 | 69 | ||
| XM_022408604.1 | XP_022264312.1 | 3 - 4 | 285 | 69 | ||
| XM_022408605.1 | XP_022264313.1 | 2 - 3 | 268 | 69 | ||
| XM_022408606.1 | XP_022264314.1 | 2 - 3 | 208 | 69 | ||
| XM_022408607.1 | XP_022264315.1 | 2 - 3 | 208 | 69 | ||
| XM_022408608.1 | XP_022264316.1 | 2 - 3 | 269 | 69 | ||
| XM_022408609.1 | XP_022264317.1 | 2 - 3 | 208 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022408605.1, XM_022408606.1, XM_022408607.1, XM_022408608.1, XM_022408609.1 transcripts.
Set Membership: |
|

LOC608697
Genomic Map