Assay Details
Gene Symbol: SPART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005635459.3 | XP_005635516.1 | 1888 | 127 | ||
XM_014107426.2 | XP_013962901.1 | 1882 | 127 | |||
XM_022409585.1 | XP_022265293.1 | 2064 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
