Assay Details
| Gene Symbol: CKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635395.3 | XP_005635452.1 | 290 | 83 | ||
| XM_005635396.3 | XP_005635453.1 | 287 | 83 | |||
| XM_005635397.3 | XP_005635454.1 | 290 | 83 | |||
| XM_005635399.3 | XP_005635456.1 | 344 | 83 | |||
| XM_022409828.1 | XP_022265536.1 | 332 | 83 | |||
| XM_022409829.1 | XP_022265537.1 | 329 | 83 | |||
| XM_022409830.1 | XP_022265538.1 | 331 | 83 | |||
| XM_022409831.1 | XP_022265539.1 | 553 | 83 | |||
| XM_022409832.1 | XP_022265540.1 | 550 | 83 | |||
| XM_022409833.1 | XP_022265541.1 | 341 | 83 | |||
| XM_022409834.1 | XP_022265542.1 | 848 | 83 | |||
More Information
Set Membership: |
|

CKAP2
Genomic Map