Assay Details
Gene Symbol: LOC477556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534751.6 | XP_534751.3 | 2 - 3 | 287 | 90 | |
GenBank mRNA | KM233847.1 | - | 2 - 3 | 245 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
