Assay Details
Gene Symbol: LOC477556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022410501.1 | XP_022266209.1 | 3 - 4 | 522 | 78 | |
XM_534751.6 | XP_534751.3 | 4 - 5 | 611 | 78 | ||
GenBank mRNA | KM233847.1 | - | 4 - 5 | 569 | 78 | |
KM233848.1 | - | 2 - 3 | 330 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
