Assay Details
| Gene Symbol: SLC25A18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637381.3 | XP_005637438.1 | 3 - 4 | 828 | 88 | |
| XM_014108382.2 | XP_013963857.1 | 5 - 6 | 1096 | 88 | ||
| XM_022411552.1 | XP_022267260.1 | 5 - 6 | 1095 | 88 | ||
| XM_022411553.1 | XP_022267261.1 | 5 - 6 | 1100 | 88 | ||
| XM_022411554.1 | XP_022267262.1 | 4 - 5 | 1002 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022411554.1 transcripts.
Set Membership: |
|

SLC25A18
Genomic Map