Assay Details
| Gene Symbol: ELOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_535104.6 | XP_535104.1 | 1 - 2 | 154 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_535104.6 transcripts.
Set Membership: |
|

ELOC
Genomic Map