Assay Details
| Gene Symbol: ELOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638049.3 | XP_005638106.1 | 3 - 4 | 312 | 107 | |
| XM_022412237.1 | XP_022267945.1 | 3 - 4 | 224 | 107 | ||
| XM_535104.6 | XP_535104.1 | 3 - 4 | 359 | 107 | ||
More Information
Set Membership: |
|

ELOC
Genomic Map