Assay Details
Gene Symbol: STN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014109067.2 | XP_013964542.1 | 2 - 3 | 512 | 106 | |
XM_014109068.2 | XP_013964543.1 | 2 - 3 | 251 | 106 | ||
XM_014109069.2 | XP_013964544.1 | 2 - 3 | 512 | 106 | ||
XM_014109070.2 | XP_013964545.1 | 2 - 3 | 512 | 106 | ||
XM_014109071.2 | XP_013964546.1 | 2 - 3 | 512 | 106 | ||
XM_014109072.2 | XP_013964547.1 | 2 - 3 | 512 | 106 | ||
XM_014109074.2 | XP_013964549.1 | 2 - 3 | 512 | 106 | ||
XM_014109075.2 | XP_013964550.1 | 2 - 3 | 512 | 106 | ||
XM_022411723.1 | XP_022267431.1 | 2 - 3 | 512 | 106 | ||
XM_022411724.1 | XP_022267432.1 | 2 - 3 | 512 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
