Assay Details
| Gene Symbol: LOC477562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014108198.1 | XP_013963673.1 | 12 - 13 | 1348 | 65 | |
| XM_014108199.2 | XP_013963674.1 | 13 - 14 | 1993 | 65 | ||
| XM_014108200.2 | XP_013963675.1 | 13 - 14 | 1701 | 65 | ||
| XM_022410769.1 | XP_022266477.1 | 14 - 15 | 1850 | 65 | ||
More Information
Set Membership: |
|

LOC477562
Genomic Map