Assay Details
Gene Symbol: LOC100856647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003640080.4 | XP_003640128.1 | 3 - 4 | 190 | 89 | |
XM_022413179.1 | XP_022268887.1 | 3 - 4 | 709 | 89 | ||
XM_022413180.1 | XP_022268888.1 | 3 - 4 | 627 | 89 | ||
XM_022413181.1 | XP_022268889.1 | 3 - 4 | 190 | 89 | ||
XM_022413182.1 | XP_022268890.1 | 3 - 4 | 190 | 89 | ||
XM_022413183.1 | XP_022268891.1 | 2 - 3 | 426 | 89 | ||
XM_022413184.1 | XP_022268892.1 | 2 - 3 | 344 | 89 |
Gene Symbol: CRYZL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022413238.1 | XP_022268946.1 | 3 - 4 | 709 | 89 | |
XM_022413239.1 | XP_022268947.1 | 3 - 4 | 627 | 89 | ||
XM_022413240.1 | XP_022268948.1 | 3 - 4 | 190 | 89 | ||
XM_022413241.1 | XP_022268949.1 | 3 - 4 | 190 | 89 | ||
XM_022413242.1 | XP_022268950.1 | 2 - 3 | 427 | 89 | ||
XM_022413243.1 | XP_022268951.1 | 2 - 3 | 345 | 89 | ||
XM_535585.6 | XP_535585.1 | 3 - 4 | 190 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022413183.1, XM_022413184.1, XM_022413242.1, XM_022413243.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
