Assay Details
| Gene Symbol: ATP5O | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638862.3 | XP_005638919.1 | 1 - 2 | 134 | 53 | |
| GenBank mRNA | CV999268.1 | - | 1 - 2 | 56 | 53 | |
| Gene Symbol: ATP5PO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638881.3 | XP_005638938.1 | 1 - 2 | 134 | 53 | |
More Information
Set Membership: |
|

ATP5O
Genomic Map