Assay Details
| Gene Symbol: MECR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617689.2 | XP_005617746.1 | 2 - 3 | 297 | 67 | |
| XM_005617690.3 | XP_005617747.1 | 2 - 3 | 227 | 67 | ||
| XM_022412444.1 | XP_022268152.1 | 2 - 3 | 223 | 67 | ||
| XM_535334.4 | XP_535334.2 | 2 - 3 | 296 | 67 | ||
More Information
Set Membership: |
|

MECR
Genomic Map