Assay Details
| Gene Symbol: COX5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287091.1 | NP_001274020.1 | 1 - 2 | 235 | 69 | |
| GenBank mRNA | CX984602.1 | - | 4 - 5 | 524 | 69 | |
| DN389481.1 | - | 1 - 2 | 170 | 69 | ||
| DN416242.1 | - | 1 - 2 | 124 | 69 | ||
| DN435073.1 | - | 1 - 2 | 192 | 69 | ||
More Information
Set Membership: |
|

COX5A
Genomic Map