Assay Details
Gene Symbol: ZNF184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014110503.2 | XP_013965978.1 | 5 - 6 | 701 | 103 | |
XM_014110504.2 | XP_013965979.1 | 5 - 6 | 568 | 103 | ||
XM_014110505.2 | XP_013965980.1 | 5 - 6 | 749 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
