Assay Details
Gene Symbol: RPP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022414491.1 | XP_022270199.1 | 4 - 5 | 467 | 104 | |
XM_022414492.1 | XP_022270200.1 | 4 - 5 | 646 | 104 | ||
XM_535875.6 | XP_535875.2 | 4 - 5 | 493 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
