Assay Details
Gene Symbol: GAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_535747.6 | XP_535747.1 | 1 - 2 | 147 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_535747.6 transcripts.
Set Membership: |
![]() ![]() ![]() |
