Assay Details
Gene Symbol: GAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_535747.6 | XP_535747.1 | 2 - 3 | 753 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_535747.6 | XP_535747.1 | 2 - 3 | 753 | 77 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |