Assay Details
| Gene Symbol: ATP5MC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_535971.6 | XP_535971.2 | 2 - 3 | 203 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_535971.6 transcripts.
Set Membership: |
|

ATP5MC3
Genomic Map