Assay Details
| Gene Symbol: ATP5MC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_535971.6 | XP_535971.2 | 4 - 5 | 481 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP5MC3| Gene Symbol: ATP5MC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_535971.6 | XP_535971.2 | 4 - 5 | 481 | 67 | |
Set Membership: |
|