Assay Details
| Gene Symbol: HMGCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638901.4 | XP_003638949.1 | 8 - 9 | 974 | 86 | |
| XM_005617813.3 | XP_005617870.1 | 8 - 9 | 956 | 86 | ||
| XM_022413394.1 | XP_022269102.1 | 9 - 10 | 991 | 86 | ||
More Information
Set Membership: |
|

HMGCL
Genomic Map