Assay Details
| Gene Symbol: ZNF131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619356.3 | XP_005619413.1 | 4 - 5 | 480 | 89 | |
| XM_005619357.3 | XP_005619414.1 | 4 - 5 | 554 | 89 | ||
| XM_005619358.3 | XP_005619415.1 | 4 - 5 | 469 | 89 | ||
| XM_014113111.2 | XP_013968586.1 | 3 - 4 | 378 | 89 | ||
| XM_862902.5 | XP_867995.1 | 4 - 5 | 479 | 89 | ||
More Information
Set Membership: |
|

ZNF131
Genomic Map