Assay Details
| Gene Symbol: ASPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116848.2 | XP_013972323.1 | 1 - 2 | 402 | 87 | |
| XM_014116849.2 | XP_013972324.1 | 1 - 2 | 402 | 87 | ||
| XM_014116850.2 | XP_013972325.1 | 1 - 2 | 401 | 87 | ||
| XM_022423478.1 | XP_022279186.1 | 3 - 4 | 558 | 87 | ||
| XM_863442.5 | XP_868535.2 | 2 - 3 | 346 | 87 | ||
More Information
Set Membership: |
|

ASPA
Genomic Map