Assay Details
| Gene Symbol: CD209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130832.1 | NP_001124304.1 | 5 - 6 | 337 | 83 | |
| XM_005632335.3 | XP_005632392.1 | 5 - 6 | 421 | 83 | ||
| XM_005632336.3 | XP_005632393.1 | 4 - 5 | 343 | 83 | ||
| XM_022406247.1 | XP_022261955.1 | 5 - 6 | 746 | 83 | ||
| GenBank mRNA | EU670674.1 | - | 5 - 6 | 337 | 83 | |
| EU670675.1 | - | 4 - 5 | 298 | 83 | ||
| EU670676.1 | - | 4 - 5 | 274 | 83 | ||
More Information
Set Membership: |
|

CD209
Genomic Map