Assay Details
| Gene Symbol: CD209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130832.1 | NP_001124304.1 | 7 - 8 | 595 | 63 | |
| XM_005632335.3 | XP_005632392.1 | 7 - 8 | 679 | 63 | ||
| XM_005632336.3 | XP_005632393.1 | 6 - 7 | 601 | 63 | ||
| XM_022406247.1 | XP_022261955.1 | 7 - 8 | 1004 | 63 | ||
| GenBank mRNA | EU670674.1 | - | 7 - 8 | 595 | 63 | |
| EU670675.1 | - | 6 - 7 | 556 | 63 | ||
| EU670676.1 | - | 6 - 7 | 532 | 63 | ||
More Information
Set Membership: |
|

CD209
Genomic Map