Assay Details
Gene Symbol: UBLCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417745.1 | XP_022273453.1 | 8 - 9 | 1015 | 136 | |
XM_546271.6 | XP_546271.2 | 9 - 10 | 1111 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
