Assay Details
| Gene Symbol: SLC5A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630253.3 | XP_005630310.1 | 16 - 17 | 2138 | 68 | |
| XM_014120384.2 | XP_013975859.1 | 16 - 17 | 2108 | 68 | ||
| XM_014120385.2 | XP_013975860.1 | 16 - 17 | 2119 | 68 | ||
| XM_014120386.2 | XP_013975861.1 | 17 - 18 | 2227 | 68 | ||
| XM_014120387.2 | XP_013975862.1 | 17 - 18 | 2197 | 68 | ||
| XM_014120388.2 | XP_013975863.1 | 17 - 18 | 2208 | 68 | ||
| XM_022404565.1 | XP_022260273.1 | 16 - 17 | 2001 | 68 | ||
More Information
Set Membership: |
|

SLC5A6
Genomic Map