Assay Details
Gene Symbol: C14H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628644.2 | XP_005628701.1 | 4 - 5 | 475 | 146 | |
XM_532494.6 | XP_532494.2 | 4 - 5 | 558 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() |
