Assay Details
Gene Symbol: C14H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628644.2 | XP_005628701.1 | 5 - 6 | 508 | 69 | |
XM_022427493.1 | XP_022283201.1 | 6 - 7 | 614 | 69 | ||
XM_532494.6 | XP_532494.2 | 5 - 6 | 591 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
