Assay Details
| Gene Symbol: MRPL42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022403311.1 | XP_022259019.1 | 4 - 5 | 357 | 64 | |
| XM_022403312.1 | XP_022259020.1 | 4 - 5 | 378 | 64 | ||
| XM_022403313.1 | XP_022259021.1 | 4 - 5 | 394 | 64 | ||
| XM_022403314.1 | XP_022259022.1 | 4 - 5 | 268 | 64 | ||
| XM_532651.6 | XP_532651.1 | 3 - 4 | 336 | 64 | ||
More Information
Set Membership: |
|

MRPL42
Genomic Map