Assay Details
Gene Symbol: GEMIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003432193.3 | XP_003432241.1 | 2 - 3 | 401 | 100 | |
XM_022404652.1 | XP_022260360.1 | 3 - 4 | 1120 | 100 | ||
XM_532941.5 | XP_532941.2 | 2 - 3 | 215 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
