Assay Details
| Gene Symbol: RHEB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022403881.1 | XP_022259589.1 | 291 | 88 | ||
| XM_532768.4 | XP_532768.1 | 291 | 88 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RHEB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022403881.1 | XP_022259589.1 | 291 | 88 | ||
| XM_532768.4 | XP_532768.1 | 291 | 88 | |||
Set Membership: |
|