Assay Details
| Gene Symbol: PRDX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_533241.6 | XP_533241.4 | 1 - 2 | 578 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PRDX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_533241.6 | XP_533241.4 | 1 - 2 | 578 | 95 | |
Set Membership: |
|