Assay Details
| Gene Symbol: DLA-DRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011723.1 | NP_001011723.1 | 2 - 3 | 326 | 73 | |
| XM_005627066.2 | XP_005627123.1 | 2 - 3 | 424 | 73 | ||
| XM_005627067.2 | XP_005627124.1 | 2 - 3 | 424 | 73 | ||
More Information
Set Membership: |
|

DLA-DRA
Genomic Map