Assay Details
Gene Symbol: C12H6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627281.2 | XP_005627338.1 | 6 - 7 | 961 | 65 | |
XM_005627282.2 | XP_005627339.1 | 7 - 8 | 1121 | 65 | ||
XM_005627283.3 | XP_005627340.1 | 5 - 6 | 923 | 65 | ||
XM_014118402.2 | XP_013973877.1 | 4 - 5 | 594 | 65 | ||
XM_022425987.1 | XP_022281695.1 | 7 - 8 | 1021 | 65 | ||
XM_022425988.1 | XP_022281696.1 | 6 - 7 | 928 | 65 | ||
XM_022425989.1 | XP_022281697.1 | 6 - 7 | 1019 | 65 | ||
XM_532127.5 | XP_532127.2 | 5 - 6 | 871 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
