Assay Details
Gene Symbol: MYCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252277.1 | NP_001239206.1 | 2 - 3 | 127 | 137 | |
GenBank mRNA | DN333656.1 | - | 2 - 3 | 127 | 137 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001252277.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
