Assay Details
Gene Symbol: NEIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630022.2 | XP_005630079.1 | 5 - 6 | 644 | 101 | |
XM_005630024.2 | XP_005630081.1 | 3 - 4 | 303 | 101 | ||
XM_022404256.1 | XP_022259964.1 | 2 - 3 | 380 | 101 | ||
XM_022404257.1 | XP_022259965.1 | 4 - 5 | 449 | 101 | ||
XM_022404258.1 | XP_022259966.1 | 4 - 5 | 477 | 101 | ||
XM_022404259.1 | XP_022259967.1 | 5 - 6 | 819 | 101 | ||
XM_022404260.1 | XP_022259968.1 | 5 - 6 | 584 | 101 | ||
XM_022404261.1 | XP_022259969.1 | 6 - 7 | 790 | 101 | ||
XM_022404263.1 | XP_022259971.1 | 5 - 6 | 714 | 101 | ||
XM_022404264.1 | XP_022259972.1 | 6 - 7 | 818 | 101 | ||
XM_022404265.1 | XP_022259973.1 | 7 - 8 | 925 | 101 | ||
XM_022404266.1 | XP_022259974.1 | 5 - 6 | 742 | 101 | ||
XM_022404267.1 | XP_022259975.1 | 5 - 6 | 461 | 101 | ||
XM_022404268.1 | XP_022259976.1 | 2 - 3 | 381 | 101 | ||
XM_022404269.1 | XP_022259977.1 | 3 - 4 | 331 | 101 | ||
XM_022404271.1 | XP_022259979.1 | 4 - 5 | 671 | 101 | ||
XM_022404272.1 | XP_022259980.1 | 4 - 5 | 429 | 101 | ||
XM_532852.5 | XP_532852.3 | 2 - 3 | 378 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005630024.2, XM_022404269.1, XM_022404271.1, XM_022404272.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
