Assay Details
| Gene Symbol: PLAAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631586.2 | XP_005631643.1 | 3 - 4 | 342 | 61 | |
| XM_533254.6 | XP_533254.3 | 3 - 4 | 342 | 61 | ||
More Information
Set Membership: |
|

PLAAT5
Genomic Map