Assay Details
| Gene Symbol: LSM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631704.2 | XP_005631761.2 | 1 | 172 | 104 | |
| Gene Symbol: LSM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_532529.5 | XP_532529.1 | 2 - 3 | 187 | 104 | |
More Information
Set Membership: |
|

LSM8
Genomic Map