Assay Details
Gene Symbol: HSD17B14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047981.1 | NP_001041446.1 | 2 - 3 | 232 | 84 | |
XM_005615748.3 | XP_005615805.1 | 2 - 3 | 336 | 84 | ||
XM_014114134.2 | XP_013969609.1 | 1 - 2 | 243 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
