Assay Details
| Gene Symbol: MRPL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022405624.1 | XP_022261332.1 | 4 - 5 | 412 | 67 | |
| XM_022405625.1 | XP_022261333.1 | 3 - 4 | 479 | 67 | ||
| XM_533221.6 | XP_533221.1 | 4 - 5 | 574 | 67 | ||
More Information
Set Membership: |
|

MRPL11
Genomic Map