Assay Details
| Gene Symbol: CYB5R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048084.1 | NP_001041549.1 | 2 - 3 | 255 | 60 | |
| GenBank mRNA | AB185964.1 | - | 2 - 3 | 255 | 60 | |
| DQ141222.1 | - | 2 - 3 | 230 | 60 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length less than or equal to 70  Recommended Assay  Canis familiaris  Probe spans exons | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 CYB5R3
CYB5R3 Genomic Map
Genomic Map Back To Top
 Back To Top