Assay Details
| Gene Symbol: CENPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628253.3 | XP_005628310.1 | 5 - 6 | 510 | 118 | |
| XM_005628254.3 | XP_005628311.1 | 5 - 6 | 512 | 118 | ||
| XM_022427025.1 | XP_022282733.1 | 5 - 6 | 480 | 118 | ||
| XM_022427026.1 | XP_022282734.1 | 5 - 6 | 445 | 118 | ||
| XM_022427027.1 | XP_022282735.1 | 4 - 5 | 415 | 118 | ||
More Information
Set Membership: |
|

CENPC
Genomic Map